The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
GenBank mRNA | AK377174.1 | - | 1 - 2 | 188 | 65 |
AK377176.1 | - | 1 - 2 | 188 | 65 | |
AK377177.1 | - | 1 - 2 | 188 | 65 | |
AK377178.1 | - | 1 - 2 | 188 | 65 | |
AK377179.1 | - | 1 - 2 | 188 | 65 | |
AK377180.1 | - | 1 - 2 | 188 | 65 | |
AK377181.1 | - | 1 - 2 | 188 | 65 | |
AK377182.1 | - | 1 - 2 | 188 | 65 | |
AK377183.1 | - | 1 - 2 | 188 | 65 | |
AK377184.1 | - | 1 - 2 | 188 | 65 |
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005694.1 | NP_005685.1 | 1 - 2 | 191 | 65 |
GenBank mRNA | AK312143.1 | - | 1 - 2 | 172 | 65 |
BC010933.1 | - | 1 - 2 | 183 | 65 | |
BC105280.1 | - | 1 - 2 | 181 | 65 | |
BC108317.1 | - | 1 - 1 | 198 | 65 | |
L77701.1 | - | 1 - 2 | 191 | 65 |