The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000476.2 | NP_000467.1 | 4 - 5 | 359 | 70 |
NM_001318121.1 | NP_001305050.1 | 4 - 5 | 346 | 70 | |
NM_001318122.1 | NP_001305051.1 | 3 - 4 | 354 | 70 | |
XM_017014427.1 | XP_016869916.1 | 4 - 5 | 1802 | 70 | |
XM_017014428.1 | XP_016869917.1 | 5 - 6 | 706 | 70 | |
GenBank mRNA | AB021871.1 | - | 2 - 3 | 206 | 70 |
AK223501.1 | - | 3 - 4 | 234 | 70 | |
AK313758.1 | - | 4 - 5 | 290 | 70 | |
BC001116.2 | - | 4 - 5 | 277 | 70 | |
BC100022.1 | - | 4 - 5 | 306 | 70 | |
BG772183.1 | - | 3 - 4 | 354 | 70 | |
BT019580.1 | - | 2 - 3 | 206 | 70 | |
CR541977.1 | - | 2 - 3 | 206 | 70 | |
CR542004.1 | - | 2 - 3 | 206 | 70 | |
CR975542.1 | - | 4 - 5 | 346 | 70 | |
DA077967.1 | - | 4 - 5 | 360 | 70 | |
EU794628.1 | - | 4 - 5 | 359 | 70 | |
HY024815.1 | - | 3 - 4 | 354 | 70 |