Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ABLIM1
Entrez Gene ID
3983
Gene Name
actin binding LIM protein 1
Gene Aliases
ABLIM, LIMAB1, LIMATIN, abLIM-1
Chromosome Location
Chr.10: 114431110 - 114821643 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001322888.1 NP_001309817.1 1 - 2 267 109
NM_001322890.1 NP_001309819.1 1 - 2 267 109
NM_001322892.1 NP_001309821.1 1 - 2 267 109
NM_001322895.1 NP_001309824.1 1 - 2 267 109
NM_001322896.1 NP_001309825.1 1 - 2 267 109
NM_001322897.1 NP_001309826.1 1 - 2 267 109
NM_001322898.1 NP_001309827.1 1 - 2 267 109
NM_001322900.1 NP_001309829.1 1 - 2 267 109
NM_006720.3 NP_006711.3 1 - 2 267 109
XM_005269826.1 XP_005269883.1 1 - 2 272 109
XM_005269827.4 XP_005269884.1 1 - 2 986 109
XM_017016258.1 XP_016871747.1 1 - 2 986 109
GenBank mRNA AK098277.1 - 1 - 2 272 109
AK298709.1 - 1 - 2 171 109
AL710059.1 - 1 - 2 272 109
BC002448.2 - 1 - 2 171 109
CR749819.1 - 1 - 2 263 109

View Full Product Details