The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_013451.3 | NP_038479.1 | 12 - 13 | 1243 | 82 |
NM_133337.2 | NP_579899.1 | 12 - 13 | 1243 | 82 | |
XM_005269693.4 | XP_005269750.1 | 12 - 13 | 1422 | 82 | |
XM_005269694.4 | XP_005269751.1 | 12 - 13 | 1422 | 82 | |
XM_011539632.2 | XP_011537934.1 | 13 - 14 | 1616 | 82 | |
XM_017016068.1 | XP_016871557.1 | 11 - 12 | 1003 | 82 | |
XM_017016069.1 | XP_016871558.1 | 11 - 12 | 1048 | 82 | |
XM_017016070.1 | XP_016871559.1 | 12 - 13 | 1430 | 82 | |
GenBank mRNA | AB033033.2 | - | 12 - 13 | 1120 | 82 |
AF182316.1 | - | 12 - 13 | 1216 | 82 | |
AF207990.1 | - | 12 - 13 | 1120 | 82 | |
BC040110.1 | - | 13 - 14 | 1433 | 82 | |
BC052617.1 | - | 12 - 13 | 1208 | 82 |