Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MYOF
Entrez Gene ID
26509
Gene Name
myoferlin
Gene Aliases
FER1L3
Chromosome Location
Chr.10: 93306429 - 93482504 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_013451.3 NP_038479.1 4 - 5 467 84
NM_133337.2 NP_579899.1 4 - 5 467 84
XM_005269693.4 XP_005269750.1 4 - 5 646 84
XM_005269694.4 XP_005269751.1 4 - 5 646 84
XM_011539632.2 XP_011537934.1 5 - 6 840 84
XM_017016068.1 XP_016871557.1 3 - 4 227 84
XM_017016069.1 XP_016871558.1 3 - 4 272 84
XM_017016070.1 XP_016871559.1 4 - 5 654 84
GenBank mRNA AB033033.2 - 4 - 5 344 84
AF182316.1 - 4 - 5 440 84
AF207990.1 - 4 - 5 344 84
BC040110.1 - 5 - 6 657 84
BC052617.1 - 4 - 5 432 84

View Full Product Details