Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SELL
Entrez Gene ID
6402
Gene Name
selectin L
Gene Aliases
CD62L, LAM1, LECAM1, LEU8, LNHR, LSEL, LYAM1, PLNHR, TQ1
Chromosome Location
Chr.1: 169690665 - 169711702 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_000655.4 NP_000646.2 7 - 8 1288 79
NR_029467.1 5 - 6 1052 79
GenBank mRNA AJ246000.1 - 7 - 8 1159 79
AK225713.1 - 7 - 8 1199 79
AK298705.1 - 5 - 6 1047 79
M25280.1 - 7 - 8 1199 79
X16070.1 - 7 - 8 1212 79
X16150.1 - 7 - 8 1175 79
X17519.1 - 7 - 8 1172 79

View Full Product Details