The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_138736.2 | NP_620073.2 | 8 - 8 | 5085 | 144 |
GenBank mRNA | AL512686.1 | - | 1 - 1 | 2794 | 144 |
BX647214.1 | - | 2 - 2 | 4002 | 144 | |
CR936770.1 | - | 8 - 8 | 4448 | 144 |