The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003635.3 | NP_003626.1 | 4 - 5 | 1895 | 59 |
XM_005270255.3 | XP_005270312.1 | 4 - 5 | 1895 | 59 | |
XM_011540310.2 | XP_011538612.1 | 3 - 4 | 2464 | 59 | |
XM_017016857.1 | XP_016872346.1 | 4 - 5 | 1895 | 59 | |
GenBank mRNA | AK303648.1 | - | 5 - 6 | 1693 | 59 |
BC035711.1 | - | 4 - 5 | 1648 | 59 | |
BC110588.1 | - | 2 - 3 | 1127 | 59 | |
BC110589.1 | - | 2 - 3 | 1127 | 59 | |
U36601.1 | - | 2 - 3 | 1118 | 59 |