The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003635.3 | NP_003626.1 | 5 - 6 | 2052 | 115 |
XM_005270255.3 | XP_005270312.1 | 5 - 6 | 2052 | 115 | |
XM_005270256.3 | XP_005270313.1 | 2 - 3 | 257 | 115 | |
XM_011540310.2 | XP_011538612.1 | 4 - 5 | 2621 | 115 | |
GenBank mRNA | AK298957.1 | - | 3 - 4 | 345 | 115 |
AK300520.1 | - | 1 - 2 | 255 | 115 | |
AK303648.1 | - | 6 - 7 | 1850 | 115 | |
BC035711.1 | - | 5 - 6 | 1805 | 115 | |
BC110588.1 | - | 3 - 4 | 1284 | 115 | |
BC110589.1 | - | 3 - 4 | 1284 | 115 | |
U36601.1 | - | 3 - 4 | 1275 | 115 |