The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003328.2 | NP_003319.2 | 7 - 8 | 669 | 152 |
XM_011513747.2 | XP_011512049.1 | 6 - 7 | 621 | 152 | |
XM_017008580.1 | XP_016864069.1 | 8 - 9 | 1871 | 152 | |
XM_017008581.1 | XP_016864070.1 | 6 - 7 | 1680 | 152 | |
GenBank mRNA | L27071.1 | - | 7 - 8 | 669 | 152 |