The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003328.2 | NP_003319.2 | 13 - 14 | 1442 | 140 |
XM_011513747.2 | XP_011512049.1 | 12 - 13 | 1394 | 140 | |
XM_011513748.2 | XP_011512050.1 | 6 - 7 | 730 | 140 | |
XM_017008580.1 | XP_016864069.1 | 14 - 15 | 2644 | 140 | |
XM_017008581.1 | XP_016864070.1 | 12 - 13 | 2453 | 140 | |
XM_017008582.1 | XP_016864071.1 | 7 - 8 | 877 | 140 | |
GenBank mRNA | AK300141.1 | - | 3 - 4 | 452 | 140 |
L27071.1 | - | 13 - 14 | 1442 | 140 |