Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TXK
Entrez Gene ID
7294
Gene Name
TXK tyrosine kinase
Gene Aliases
BTKL, PSCTK5, PTK4, RLK, TKL
Chromosome Location
Chr.4: 48066393 - 48135322 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_003328.2 NP_003319.2 10 - 11 1040 89
XM_011513747.2 XP_011512049.1 9 - 10 992 89
XM_011513748.2 XP_011512050.1 3 - 4 328 89
XM_017008580.1 XP_016864069.1 11 - 12 2242 89
XM_017008581.1 XP_016864070.1 9 - 10 2051 89
XM_017008582.1 XP_016864071.1 4 - 5 475 89
GenBank mRNA L27071.1 - 10 - 11 1040 89

View Full Product Details