The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001256468.1 | NP_001243397.1 | 8 - 9 | 2138 | 111 |
XM_017016005.1 | XP_016871494.1 | 8 - 9 | 2047 | 111 | |
GenBank mRNA | AF119230.1 | - | 7 - 8 | 1752 | 111 |