The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004439.7 | NP_004430.4 | 7 - 8 | 2439 | 78 |
NM_182472.4 | NP_872272.2 | 7 - 8 | 2439 | 78 | |
XM_017007879.1 | XP_016863368.1 | 6 - 7 | 2446 | 78 | |
XM_017007881.1 | XP_016863370.1 | 5 - 6 | 2290 | 78 | |
GenBank mRNA | L36644.1 | - | 7 - 8 | 1614 | 78 |