The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003601.3 | NP_003592.3 | 14 - 15 | 2360 | 128 |
GenBank mRNA | AB010882.1 | - | 14 - 15 | 2100 | 128 |
AK302841.1 | - | 15 - 16 | 1995 | 128 | |
AK316444.1 | - | 15 - 16 | 2162 | 128 | |
BC023144.1 | - | 14 - 15 | 2111 | 128 |