Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SMARCA5
Entrez Gene ID
8467
Gene Name
SWI/SNF related, matrix associated, actin dependent regulator of chromatin, subfamily a, member 5
Gene Aliases
ISWI, SNF2H, WCRF135, hISWI, hSNF2H
Chromosome Location
Chr.4: 143513463 - 143557489 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_003601.3 NP_003592.3 17 - 18 2743 77
GenBank mRNA AB010882.1 - 17 - 18 2483 77
AK302841.1 - 18 - 19 2378 77
AK316444.1 - 18 - 19 2545 77
BC023144.1 - 17 - 18 2494 77

View Full Product Details