The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003601.3 | NP_003592.3 | 8 - 9 | 1544 | 96 |
GenBank mRNA | AB010882.1 | - | 8 - 9 | 1284 | 96 |
AK302841.1 | - | 9 - 10 | 1179 | 96 | |
AK308424.1 | - | 8 - 9 | 1274 | 96 | |
AK316444.1 | - | 9 - 10 | 1346 | 96 | |
BC023144.1 | - | 8 - 9 | 1295 | 96 |