The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001256421.1 | NP_001243350.1 | 2 - 3 | 360 | 153 |
NM_001256422.1 | NP_001243351.1 | 2 - 3 | 364 | 153 | |
NM_032717.4 | NP_116106.2 | 1 - 2 | 447 | 153 | |
XM_017008780.1 | XP_016864269.1 | 2 - 3 | 516 | 153 | |
XM_017008781.1 | XP_016864270.1 | 2 - 3 | 523 | 153 | |
GenBank mRNA | AK055749.1 | - | 2 - 3 | 359 | 153 |
AK223543.1 | - | 2 - 3 | 331 | 153 | |
AY358100.1 | - | 1 - 2 | 264 | 153 | |
BC090956.1 | - | 1 - 2 | 353 | 153 | |
DA393801.1 | - | 2 - 3 | 364 | 153 | |
DA673378.1 | - | 1 - 2 | 447 | 153 | |
DQ324782.1 | - | 1 - 2 | 144 | 153 | |
DQ345298.1 | - | 1 - 2 | 444 | 153 |