The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001256421.1 | NP_001243350.1 | 6 - 7 | 865 | 78 |
NM_001256422.1 | NP_001243351.1 | 6 - 7 | 869 | 78 | |
NM_032717.4 | NP_116106.2 | 5 - 6 | 952 | 78 | |
XM_011532383.1 | XP_011530685.1 | 4 - 5 | 758 | 78 | |
XM_011532384.2 | XP_011530686.1 | 6 - 7 | 2953 | 78 | |
XM_017008780.1 | XP_016864269.1 | 6 - 7 | 1021 | 78 | |
XM_017008781.1 | XP_016864270.1 | 6 - 7 | 1028 | 78 | |
GenBank mRNA | AB075872.1 | - | 2 - 3 | 150 | 78 |
AK223543.1 | - | 6 - 7 | 836 | 78 | |
AY358100.1 | - | 5 - 6 | 769 | 78 | |
BC090956.1 | - | 5 - 6 | 858 | 78 | |
DQ324782.1 | - | 5 - 6 | 649 | 78 | |
DQ345298.1 | - | 5 - 6 | 949 | 78 |