Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MNAT1
Entrez Gene ID
4331
Gene Name
MNAT1, CDK activating kinase assembly factor
Gene Aliases
CAP35, MAT1, RNF66, TFB3
Chromosome Location
Chr.14: 60734735 - 60969881 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001177963.1 NP_001171434.1 5 - 6 681 139

View Full Product Details