Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FNBP1L
Entrez Gene ID
54874
Gene Name
formin binding protein 1 like
Gene Aliases
C1orf39, TOCA1
Chromosome Location
Chr.1: 93448067 - 93569671 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001164473.2 NP_001157945.1 9 - 10 1145 107
GenBank mRNA AK299760.1 - 4 - 5 531 107
AK299943.1 - 3 - 4 441 107
AK315981.1 - 1 - 2 155 107

View Full Product Details