The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_018945.3 | NP_061818.1 | 1 - 2 | 330 | 109 |
GenBank mRNA | AB038040.1 | - | 1 - 2 | 332 | 109 |
AJ251860.1 | - | 1 - 2 | 324 | 109 | |
AK290582.1 | - | 1 - 2 | 363 | 109 | |
BC070267.1 | - | 1 - 2 | 262 | 109 | |
BC075082.2 | - | 1 - 2 | 53 | 109 | |
BC075083.2 | - | 1 - 2 | 57 | 109 |