The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_018945.3 | NP_061818.1 | 7 - 8 | 883 | 76 |
XM_005266931.3 | XP_005266988.1 | 6 - 7 | 3732 | 76 | |
XM_011535755.2 | XP_011534057.1 | 8 - 9 | 712 | 76 | |
XM_017010741.1 | XP_016866230.1 | 6 - 7 | 1518 | 76 | |
GenBank mRNA | AB038040.1 | - | 7 - 8 | 885 | 76 |
AB209990.1 | - | 6 - 7 | 989 | 76 | |
AJ251860.1 | - | 7 - 8 | 877 | 76 | |
AK290582.1 | - | 7 - 8 | 916 | 76 | |
BC075082.2 | - | 7 - 8 | 606 | 76 | |
BC075083.2 | - | 7 - 8 | 610 | 76 | |
EF108327.1 | - | 6 - 7 | 991 | 76 |