The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_006208.2 | NP_006199.2 | 11 - 12 | 1179 | 125 |
XM_011535896.1 | XP_011534198.1 | 2 - 3 | 108 | 99 | |
GenBank mRNA | AK314870.1 | - | 11 - 12 | 1006 | 125 |
BC059375.1 | - | 11 - 12 | 1179 | 125 | |
BX537580.1 | - | 3 - 4 | 203 | 99 | |
D12485.1 | - | 11 - 12 | 1175 | 125 | |
M57736.1 | - | 11 - 12 | 1166 | 125 |