Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ENPP1
Entrez Gene ID
5167
Gene Name
ectonucleotide pyrophosphatase/phosphodiesterase 1
Gene Aliases
ARHR2, COLED, M6S1, NPP1, NPPS, PC-1, PCA1, PDNP1
Chromosome Location
Chr.6: 131808016 - 131895155 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_006208.2 NP_006199.2 6 - 7 732 151
GenBank mRNA AK314870.1 - 6 - 7 559 151
BC059375.1 - 6 - 7 732 151
D12485.1 - 6 - 7 728 151
M57736.1 - 6 - 7 719 151

View Full Product Details