The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | XM_005270590.2 | XP_005270647.1 | 24 - 25 | 2816 | 100 |
XM_005270591.2 | XP_005270648.1 | 24 - 25 | 2798 | 100 | |
XM_005270593.2 | XP_005270650.1 | 20 - 21 | 2552 | 100 | |
XM_006710423.2 | XP_006710486.1 | 23 - 24 | 2690 | 100 | |
XM_017000559.1 | XP_016856048.1 | 24 - 25 | 2728 | 100 | |
GenBank mRNA | BC007912.2 | - | 12 - 13 | 1178 | 100 |
BC039308.1 | - | 23 - 24 | 3644 | 100 |