Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MACF1
Entrez Gene ID
23499
Gene Name
microtubule-actin crosslinking factor 1
Gene Aliases
ABP620, ACF7, MACF, OFC4
Chromosome Location
Chr.1: 39084167 - 39487138 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_012090.5 NP_036222.3 30 - 31 4098 140
XM_005270694.1 XP_005270751.1 26 - 27 4759 140
XM_005270695.1 XP_005270752.1 26 - 27 4759 140
XM_005270696.1 XP_005270753.1 26 - 27 4759 140
XM_005270697.1 XP_005270754.1 26 - 27 4759 140
XM_005270698.1 XP_005270755.1 26 - 27 4759 140
XM_006710519.1 XP_006710582.1 26 - 27 4759 140
XM_006710520.2 XP_006710583.1 26 - 27 4759 140
XM_006710521.1 XP_006710584.1 26 - 27 4759 140
XM_006710522.1 XP_006710585.1 26 - 27 4759 140
XM_006710524.2 XP_006710587.1 26 - 27 4759 140
XM_006710526.3 XP_006710589.1 31 - 32 4191 140
XM_006710528.3 XP_006710591.1 31 - 32 4752 140
XM_006710529.3 XP_006710592.1 31 - 32 4109 140
XM_006710530.3 XP_006710593.1 31 - 32 4997 140
XM_006710531.3 XP_006710594.1 31 - 32 4728 140
XM_006710532.3 XP_006710595.1 30 - 31 4656 140
XM_006710533.2 XP_006710596.1 30 - 31 4656 140
XM_006710535.3 XP_006710598.1 31 - 32 4261 140
XM_006710536.2 XP_006710599.1 26 - 27 4759 140
XM_006710538.2 XP_006710601.1 26 - 27 4759 140
XM_011541141.2 XP_011539443.1 26 - 27 4759 140
XM_011541142.2 XP_011539444.1 26 - 27 4759 140
XM_017000873.1 XP_016856362.1 26 - 27 4759 140
XM_017000874.1 XP_016856363.1 26 - 27 4759 140
XM_017000875.1 XP_016856364.1 30 - 31 4081 140
XM_017000876.1 XP_016856365.1 30 - 31 4165 140
XM_017000877.1 XP_016856366.1 31 - 32 4087 140
XM_017000878.1 XP_016856367.1 31 - 32 3950 140
XM_017000880.1 XP_016856369.1 26 - 27 4759 140
XM_017000881.1 XP_016856370.1 30 - 31 4073 140
XM_017000882.1 XP_016856371.1 26 - 27 4759 140
GenBank mRNA AB007934.3 - 10 - 11 1446 140
AB029290.1 - 30 - 31 4097 140
AB033077.1 - 10 - 11 1446 140
AK127519.1 - 26 - 27 4610 140

View Full Product Details