Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MACF1
Entrez Gene ID
23499
Gene Name
microtubule-actin crosslinking factor 1
Gene Aliases
ABP620, ACF7, MACF, OFC4
Chromosome Location
Chr.1: 39084167 - 39487138 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_012090.5 NP_036222.3 76 - 77 13633 92
XM_005270694.1 XP_005270751.1 77 - 78 20495 92
XM_005270695.1 XP_005270752.1 77 - 78 20495 92
XM_005270696.1 XP_005270753.1 77 - 78 20495 92
XM_005270697.1 XP_005270754.1 77 - 78 20495 92
XM_005270698.1 XP_005270755.1 77 - 78 20495 92
XM_006710519.1 XP_006710582.1 76 - 77 20486 92
XM_006710520.2 XP_006710583.1 76 - 77 20432 92
XM_006710521.1 XP_006710584.1 77 - 78 20495 92
XM_006710522.1 XP_006710585.1 77 - 78 20495 92
XM_006710524.2 XP_006710587.1 76 - 77 20261 92
XM_006710526.3 XP_006710589.1 82 - 83 19927 92
XM_006710528.3 XP_006710591.1 82 - 83 20488 92
XM_006710529.3 XP_006710592.1 82 - 83 19845 92
XM_006710530.3 XP_006710593.1 82 - 83 20733 92
XM_006710531.3 XP_006710594.1 82 - 83 20464 92
XM_006710532.3 XP_006710595.1 81 - 82 20392 92
XM_006710533.2 XP_006710596.1 81 - 82 20392 92
XM_006710535.3 XP_006710598.1 82 - 83 19997 92
XM_006710536.2 XP_006710599.1 72 - 73 14294 92
XM_011541141.2 XP_011539443.1 77 - 78 20441 92
XM_011541142.2 XP_011539444.1 76 - 77 15044 92
XM_017000873.1 XP_016856362.1 76 - 77 20486 92
XM_017000874.1 XP_016856363.1 76 - 77 20486 92
XM_017000875.1 XP_016856364.1 81 - 82 19817 92
XM_017000876.1 XP_016856365.1 81 - 82 19901 92
XM_017000877.1 XP_016856366.1 82 - 83 19823 92
XM_017000878.1 XP_016856367.1 82 - 83 19686 92
XM_017000879.1 XP_016856368.1 47 - 48 15257 92
XM_017000880.1 XP_016856369.1 71 - 72 14231 92
XM_017000881.1 XP_016856370.1 76 - 77 13608 92
XM_017000882.1 XP_016856371.1 71 - 72 14231 92
GenBank mRNA AB007934.3 - 54 - 55 10591 92
AB029290.1 - 76 - 77 13632 92
AF317696.1 - 46 - 47 15207 92

View Full Product Details