Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
C12orf42
Entrez Gene ID
374470
Gene Name
chromosome 12 open reading frame 42
Gene Aliases
-
Chromosome Location
Chr.12: 103047669 - 103525258 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001099336.2 NP_001092806.1 4 - 5 523 121
NM_198521.3 NP_940923.2 4 - 5 388 121
XM_006719377.3 XP_006719440.1 3 - 4 326 121
XM_011538291.1 XP_011536593.1 4 - 5 383 121
XM_011538292.2 XP_011536594.1 4 - 5 352 121
XM_011538293.2 XP_011536595.1 4 - 5 359 121
XM_011538294.2 XP_011536596.1 4 - 5 333 121
XM_011538295.1 XP_011536597.1 4 - 5 415 121
XM_011538297.2 XP_011536599.1 5 - 6 1399 121
XM_011538300.1 XP_011536602.1 5 - 6 568 121
XM_011538305.1 XP_011536607.1 4 - 5 585 121
XM_017019276.1 XP_016874765.1 4 - 5 477 121
XM_017019277.1 XP_016874766.1 7 - 8 852 121
XM_017019278.1 XP_016874767.1 6 - 7 631 121
XM_017019279.1 XP_016874768.1 7 - 8 1157 121
XM_017019280.1 XP_016874769.1 7 - 8 857 121
GenBank mRNA AK058052.1 - 4 - 5 346 121
BC117392.1 - 4 - 5 346 121
BC117394.1 - 4 - 5 346 121

View Full Product Details