The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001099336.2 | NP_001092806.1 | 5 - 6 | 894 | 98 |
NM_001278419.1 | NP_001265348.1 | 6 - 7 | 989 | 98 | |
NM_001278420.1 | NP_001265349.1 | 6 - 7 | 854 | 98 | |
NM_198521.3 | NP_940923.2 | 5 - 6 | 759 | 98 | |
XM_006719377.3 | XP_006719440.1 | 4 - 5 | 697 | 98 | |
XM_011538291.1 | XP_011536593.1 | 5 - 6 | 754 | 98 | |
XM_011538292.2 | XP_011536594.1 | 5 - 6 | 723 | 98 | |
XM_011538293.2 | XP_011536595.1 | 5 - 6 | 730 | 98 | |
XM_011538294.2 | XP_011536596.1 | 5 - 6 | 704 | 98 | |
XM_011538295.1 | XP_011536597.1 | 5 - 6 | 786 | 98 | |
XM_011538297.2 | XP_011536599.1 | 6 - 7 | 1770 | 98 | |
XM_011538300.1 | XP_011536602.1 | 6 - 7 | 939 | 98 | |
XM_011538305.1 | XP_011536607.1 | 5 - 6 | 956 | 98 | |
XM_017019276.1 | XP_016874765.1 | 5 - 6 | 848 | 98 | |
XM_017019277.1 | XP_016874766.1 | 8 - 9 | 1223 | 98 | |
XM_017019278.1 | XP_016874767.1 | 7 - 8 | 1002 | 98 | |
XM_017019279.1 | XP_016874768.1 | 8 - 9 | 1528 | 98 | |
XM_017019280.1 | XP_016874769.1 | 8 - 9 | 1228 | 98 | |
GenBank mRNA | AK058052.1 | - | 5 - 6 | 717 | 98 |
BC044617.1 | - | 6 - 7 | 950 | 98 | |
BC117392.1 | - | 5 - 6 | 717 | 98 | |
BC117394.1 | - | 5 - 6 | 717 | 98 |