Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SLCO6A1
Entrez Gene ID
133482
Gene Name
solute carrier organic anion transporter family member 6A1
Gene Aliases
CT48, GST, OATP-I, OATP6A1, OATPY
Chromosome Location
Chr.5: 102371782 - 102502213 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_011543150.1 XP_011541452.1 1 - 2 94 147

View Full Product Details