Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SDF2
Entrez Gene ID
6388
Gene Name
stromal cell derived factor 2
Gene Aliases
-
Chromosome Location
Chr.17: 28648356 - 28662189 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_011525106.2 XP_011523408.1 3 - 4 503 104

View Full Product Details