Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
KCND2
Entrez Gene ID
3751
Gene Name
potassium voltage-gated channel subfamily D member 2
Gene Aliases
KV4.2, RK5
Chromosome Location
Chr.7: 120273635 - 120750333 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_012281.2 NP_036413.1 3 - 4 2339 70
GenBank mRNA AB028967.1 - 3 - 4 2339 70
AJ010969.1 - 3 - 4 1803 70
BC110449.1 - 3 - 4 1993 70
BC110450.1 - 3 - 4 1993 70

View Full Product Details