The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001144877.2 | NP_001138349.1 | 6 - 7 | 635 | 155 |
NM_173690.4 | NP_775961.2 | 7 - 8 | 704 | 155 | |
GenBank mRNA | AK093983.1 | - | 6 - 7 | 594 | 155 |
AK098024.1 | - | 7 - 8 | 641 | 155 | |
BC104030.2 | - | 7 - 8 | 632 | 155 | |
BC104031.1 | - | 6 - 7 | 563 | 155 | |
BC104032.1 | - | 6 - 7 | 563 | 155 |