Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SDE2
Entrez Gene ID
163859
Gene Name
SDE2 telomere maintenance homolog
Gene Aliases
C1orf55, dJ671D7.1
Chromosome Location
Chr.1: 225982702 - 225999365 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_152608.3 NP_689821.3 6 - 7 1182 80
GenBank mRNA AK092701.1 - 6 - 7 1008 80
AK127780.1 - 5 - 6 1112 80
AK291008.1 - 6 - 7 1145 80
BC071563.1 - 6 - 7 1160 80

View Full Product Details