Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DNA2
Entrez Gene ID
1763
Gene Name
DNA replication helicase/nuclease 2
Gene Aliases
DNA2L, hDNA2
Chromosome Location
Chr.10: 68414064 - 68472521 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001080449.2 NP_001073918.2 7 - 8 1159 99
NR_102264.1 8 - 9 1139 99
XM_006717680.3 XP_006717743.1 8 - 9 1539 99
XM_017015799.1 XP_016871288.1 4 - 5 548 99
GenBank mRNA AK308333.1 - 7 - 8 1159 99
AK308358.1 - 5 - 6 800 99
BC063664.1 - 7 - 8 1072 99
BC111740.1 - 8 - 9 1139 99
D42046.1 - 7 - 8 1101 99

View Full Product Details