The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001012426.1 | NP_001012426.1 | 2 - 3 | 665 | 112 |
NM_001012427.1 | NP_001012427.1 | 2 - 3 | 665 | 112 | |
NM_138457.2 | NP_612466.1 | 2 - 3 | 665 | 112 | |
XM_006714991.3 | XP_006715054.1 | 2 - 3 | 470 | 112 | |
XM_011514289.2 | XP_011512591.1 | 2 - 3 | 542 | 112 | |
XM_011514290.2 | XP_011512592.1 | 2 - 3 | 543 | 112 | |
XM_011514291.2 | XP_011512593.1 | 2 - 3 | 541 | 112 | |
XM_017010233.1 | XP_016865722.1 | 2 - 3 | 1674 | 112 | |
XM_017010234.1 | XP_016865723.1 | 2 - 3 | 542 | 112 | |
GenBank mRNA | AB080747.1 | - | 1 - 2 | 219 | 112 |
BC040962.1 | - | 2 - 3 | 665 | 112 | |
BC052803.1 | - | 2 - 3 | 443 | 112 |