The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001012426.1 | NP_001012426.1 | 7 - 8 | 1325 | 156 |
NM_001012427.1 | NP_001012427.1 | 7 - 8 | 1319 | 156 | |
NM_138457.2 | NP_612466.1 | 7 - 8 | 1322 | 156 | |
XM_006714991.3 | XP_006715054.1 | 7 - 8 | 1127 | 156 | |
XM_011514289.2 | XP_011512591.1 | 7 - 8 | 1223 | 156 | |
XM_011514290.2 | XP_011512592.1 | 7 - 8 | 1218 | 156 | |
XM_011514291.2 | XP_011512593.1 | 7 - 8 | 1222 | 156 | |
XM_011514292.2 | XP_011512594.1 | 6 - 7 | 1128 | 156 | |
XM_011514293.2 | XP_011512595.1 | 5 - 6 | 744 | 156 | |
XM_017010233.1 | XP_016865722.1 | 7 - 8 | 2355 | 156 | |
XM_017010234.1 | XP_016865723.1 | 7 - 8 | 1223 | 156 | |
GenBank mRNA | AB080747.1 | - | 6 - 7 | 879 | 156 |
BC040962.1 | - | 7 - 8 | 1322 | 156 | |
BC052803.1 | - | 7 - 8 | 1097 | 156 |