Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CA9
Entrez Gene ID
768
Gene Name
carbonic anhydrase 9
Gene Aliases
CAIX, MN
Chromosome Location
Chr.9: 35673859 - 35681159 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001216.2 NP_001207.2 10 - 11 1355 67
GenBank mRNA AJ010588.1 - 10 - 11 1322 67
BC014950.1 - 10 - 11 1417 67
EF122496.1 - 8 - 9 1177 67
KU177952.1 - 10 - 11 1313 67
KU177953.1 - 10 - 11 1426 67
X66839.1 - 10 - 11 1355 67

View Full Product Details