The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001216.2 | NP_001207.2 | 3 - 4 | 642 | 70 |
GenBank mRNA | AJ010588.1 | - | 3 - 4 | 609 | 70 |
AL554705.3 | - | 3 - 4 | 641 | 70 | |
BC014950.1 | - | 3 - 4 | 704 | 70 | |
EF122496.1 | - | 3 - 4 | 642 | 70 | |
KU177952.1 | - | 3 - 4 | 600 | 70 | |
KU177953.1 | - | 3 - 4 | 600 | 70 | |
X66839.1 | - | 3 - 4 | 642 | 70 |