Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SPIRE1
Entrez Gene ID
56907
Gene Name
spire type actin nucleation factor 1
Gene Aliases
Spir-1
Chromosome Location
Chr.18: 12446512 - 12658102 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001128627.1 NP_001122099.1 8 - 9 935 93
NM_020148.2 NP_064533.3 8 - 9 1237 93
XM_005258122.4 XP_005258179.1 8 - 9 1420 93
GenBank mRNA AJ277587.1 - 6 - 7 824 93
AK290180.1 - 6 - 7 795 93
AK301711.1 - 7 - 8 704 93
AL833817.1 - 9 - 10 915 93
BC115005.1 - 8 - 9 1062 93
BC125207.1 - 6 - 7 751 93

View Full Product Details