The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_016551.2 | NP_057635.1 | 1 - 2 | 332 | 86 |
XM_005253391.3 | XP_005253448.1 | 1 - 2 | 353 | 86 | |
XM_011520691.1 | XP_011518993.1 | 1 - 2 | 353 | 86 | |
XM_017019463.1 | XP_016874952.1 | 1 - 2 | 353 | 86 | |
GenBank mRNA | AB032470.1 | - | 1 - 2 | 131 | 86 |
AK002031.1 | - | 1 - 2 | 98 | 86 | |
AK023085.1 | - | 1 - 2 | 320 | 86 | |
BC005176.2 | - | 1 - 2 | 257 | 86 |