The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_016551.2 | NP_057635.1 | 5 - 6 | 932 | 81 |
XM_005253391.3 | XP_005253448.1 | 5 - 6 | 953 | 81 | |
XM_011520690.2 | XP_011518992.1 | 4 - 5 | 795 | 81 | |
XM_017019463.1 | XP_016874952.1 | 5 - 6 | 1043 | 81 | |
XM_017019464.1 | XP_016874953.1 | 3 - 4 | 605 | 81 | |
XM_017019465.1 | XP_016874954.1 | 3 - 4 | 605 | 81 | |
XM_017019466.1 | XP_016874955.1 | 4 - 5 | 772 | 81 | |
GenBank mRNA | AB032470.1 | - | 5 - 6 | 731 | 81 |
AK002031.1 | - | 5 - 6 | 698 | 81 | |
AK023085.1 | - | 5 - 6 | 920 | 81 | |
BC005176.2 | - | 5 - 6 | 857 | 81 |