The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014044.5 | NP_054763.2 | 1 - 2 | 147 | 92 |
XM_006712404.2 | XP_006712467.1 | 1 - 2 | 157 | 92 | |
GenBank mRNA | AF077038.1 | - | 1 - 2 | 99 | 92 |
AK222599.1 | - | 1 - 2 | 113 | 92 | |
AL080115.1 | - | 1 - 2 | 151 | 92 | |
AY017215.1 | - | 1 - 2 | 115 | 92 | |
BC091481.1 | - | 1 - 2 | 137 | 92 |