The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004333.4 | NP_004324.2 | 8 - 9 | 1199 | 85 |
XM_005250045.2 | XP_005250102.1 | 8 - 9 | 1146 | 85 | |
GenBank mRNA | AK314346.1 | - | 8 - 9 | 1146 | 85 |
BC101757.1 | - | 8 - 9 | 1160 | 85 | |
BC112079.1 | - | 8 - 9 | 1160 | 85 | |
M95712.2 | - | 8 - 9 | 1199 | 85 |