The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_018290.3 | NP_060760.2 | 6 - 7 | 794 | 88 |
XM_011513711.2 | XP_011512013.1 | 7 - 8 | 897 | 88 | |
GenBank mRNA | AF109360.1 | - | 6 - 7 | 734 | 88 |
AK001845.1 | - | 6 - 7 | 737 | 88 | |
AK223237.1 | - | 6 - 7 | 736 | 88 | |
AK297752.1 | - | 4 - 5 | 531 | 88 | |
AK303374.1 | - | 7 - 8 | 902 | 88 | |
AL136705.1 | - | 6 - 7 | 751 | 88 | |
BC010087.2 | - | 6 - 7 | 734 | 88 | |
CR457274.1 | - | 6 - 7 | 721 | 88 |