The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001144881.1 | NP_001138353.1 | 1 - 2 | 242 | 119 |
GenBank mRNA | AK122877.1 | - | 1 - 2 | 243 | 119 |