The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001105558.1 | NP_001099028.1 | 5 - 6 | 1282 | 104 |
GenBank mRNA | AK131218.1 | - | 5 - 6 | 1282 | 104 |
BM790836.1 | - | 2 - 3 | 201 | 104 |