The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001013694.2 | NP_001013716.2 | 2 - 3 | 255 | 144 |
XM_017028799.1 | XP_016884288.1 | 2 - 3 | 267 | 144 | |
GenBank mRNA | AK293916.1 | - | 3 - 4 | 235 | 144 |
BC066962.1 | - | 3 - 4 | 238 | 144 | |
DA325985.1 | - | 2 - 3 | 250 | 144 |