Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FAM98C
Entrez Gene ID
147965
Gene Name
family with sequence similarity 98 member C
Gene Aliases
-
Chromosome Location
Chr.19: 38403124 - 38409088 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_174905.3 NP_777565.3 7 - 8 103
GenBank mRNA BC117258.1 - 7 - 8 103
BC143830.1 - 7 - 8 103
BC171747.1 - 4 - 5 103
BI871449.1 - 4 - 5 103
BX641942.1 - 1 - 2 356 103

View Full Product Details